MilliporeSigma
  • Inhibition of protein synthesis by the T cell receptor-inducible human TDAG51 gene product.
Inhibition of protein synthesis by the T cell receptor-inducible human TDAG51 gene product.
Cellular signalling (2001-05-23)
T Hinz, S Flindt, A Marx, O Janssen, D Kabelitz
ABSTRACT
The T cell death associated gene 51 (TDAG51) was shown to be required for T cell receptor (TCR)-dependent induction of Fas/Apo1/CD95 expression in a murine T cell hybridoma. Despite the absence of a nuclear localization sequence and a nucleic acid binding domain, it was suggested to be localized in the nucleus and to function as a transcription factor regulating Fas-expression. However, we demonstrate that the human (h)TDAG51 protein is localized in the cytoplasm and the nucleoli, suggesting a role in ribosome biogenesis and/or translation regulation. Indeed, it strongly inhibited translation of a luciferase mRNA in a reticulocyte translational extract. Furthermore, cotransfection of hTDAG51 and the luciferase gene into 293T cells resulted in a strong inhibition of luciferase mRNA translation. Our findings were further strengthened by isolating in a yeast two-hybrid screen three proteins which are involved in the regulation of translation. We speculate that hTDAG51 couples TCR signaling to inhibition of protein biosynthesis in activated T lymphocytes.