MilliporeSigma
  • Molecular cloning and characterization of UDP-GlcNAc:lactosylceramide beta 1,3-N-acetylglucosaminyltransferase (beta 3Gn-T5), an essential enzyme for the expression of HNK-1 and Lewis X epitopes on glycolipids.
Molecular cloning and characterization of UDP-GlcNAc:lactosylceramide beta 1,3-N-acetylglucosaminyltransferase (beta 3Gn-T5), an essential enzyme for the expression of HNK-1 and Lewis X epitopes on glycolipids.
The Journal of biological chemistry (2001-04-03)
A Togayachi, T Akashima, R Ookubo, T Kudo, S Nishihara, H Iwasaki, A Natsume, H Mio, J Inokuchi, T Irimura, K Sasaki, H Narimatsu
ABSTRACT
A new member of the UDP-N-acetylglucosamine:beta-galactose beta1,3-N-acetylglucosaminyltransferase (beta3Gn-T) family having the beta3Gn-T motifs was cloned from rat and human cDNA libraries and named beta3Gn-T5 based on its position in a phylogenetic tree. We concluded that beta3Gn-T5 is the most feasible candidate for lactotriaosylceramide (Lc(3)Cer) synthase, an important enzyme which plays a key role in the synthesis of lacto- or neolacto-series carbohydrate chains on glycolipids. beta3Gn-T5 exhibited strong activity to transfer GlcNAc to glycolipid substrates, such as lactosylceramide (LacCer) and neolactotetraosylceramide (nLc(4)Cer; paragloboside), resulting in the synthesis of Lc(3)Cer and neolactopentaosylceramide (nLc(5)Cer), respectively. A marked decrease in LacCer and increase in nLc(4)Cer was detected in Namalwa cells stably expressing beta3Gn-T5. This indicated that beta3Gn-T5 exerted activity to synthesize Lc(3)Cer and decrease LacCer, followed by conversion to nLc(4)Cer via endogenous galactosylation. The following four findings further supported that beta3Gn-T5 is Lc(3)Cer synthase. 1) The beta3Gn-T5 transcript levels in various cells were consistent with the activity levels of Lc(3)Cer synthase in those cells. 2) The beta3Gn-T5 transcript was presented in various tissues and cultured cells. 3) The beta3Gn-T5 expression was up-regulated by stimulation with retinoic acid and down-regulated with 12-O-tetradecanoylphorbol-13-acetate in HL-60 cells. 4) The changes in beta3Gn-T5 transcript levels during the rat brain development were determined. Points 2, 3, and 4 were consistent with the Lc(3)Cer synthase activity reported previously.