MilliporeSigma
  • Human immunodeficiency virus-1 Tat activates NF-κB via physical interaction with IκB-α and p65.
Human immunodeficiency virus-1 Tat activates NF-κB via physical interaction with IκB-α and p65.
Nucleic acids research (2011-12-22)
Giuseppe Fiume, Eleonora Vecchio, Annamaria De Laurentiis, Francesca Trimboli, Camillo Palmieri, Antonio Pisano, Cristina Falcone, Marilena Pontoriero, Annalisa Rossi, Annarita Scialdone, Francesca Fasanella Masci, Giuseppe Scala, Ileana Quinto
ABSTRACT
Nuclear factor (NF)-κB is a master regulator of pro-inflammatory genes and is upregulated in human immunodeficiency virus 1 (HIV-1) infection. Mechanisms underlying the NF-κB deregulation by HIV-1 are relevant for immune dysfunction in AIDS. We report that in single round HIV-1 infection, or single-pulse PMA stimulation, the HIV-1 Tat transactivator activated NF-κB by hijacking the inhibitor IκB-α and by preventing the repressor binding to the NF-κB complex. Moreover, Tat associated with the p65 subunit of NF-κB and increased the p65 DNA-binding affinity and transcriptional activity. The arginine- and cysteine-rich domains of Tat were required for IκB-α and p65 association, respectively, and for sustaining the NF-κB activity. Among an array of NF-κB-responsive genes, Tat mostly activated the MIP-1α expression in a p65-dependent manner, and bound to the MIP-1α NF-κB enhancer thus promoting the recruitment of p65 with displacement of IκB-α; similar findings were obtained for the NF-κB-responsive genes CSF3, LTA, NFKBIA and TLR2. Our results support a novel mechanism of NF-κB activation via physical interaction of Tat with IκB-α and p65, and may contribute to further insights into the deregulation of the inflammatory response by HIV-1.