MilliporeSigma
  • Association of rs1219648 in FGFR2 and rs1042522 in TP53 with premenopausal breast cancer in an Iranian Azeri population.
Association of rs1219648 in FGFR2 and rs1042522 in TP53 with premenopausal breast cancer in an Iranian Azeri population.
Asian Pacific journal of cancer prevention : APJCP (2014-10-09)
Zahra Saadatian, Jalal Gharesouran, Morteza Ghojazadeh, Sahar Ghohari-Lasaki, Najime Tarkesh-Esfahani, Seyyed Mojtaba Mohaddes Ardebili
ABSTRACT
Breast cancer is the most common cancer among women in the world. In Iran, the incidence of breast cancer is on the increase. We here studied the association of rs1219648 in FGFR2 and rs1042522 in TP53 and their interaction in development of early onset sporadic breast cancer in Iranian Azeri population to evaluate epistatic effects on the risk of mammary neoplasia. We genotyped the two polymorphisms in 100 women with early onset breast cancer and 100 healthy women by PCR-RFLP. Allele frequency differences were tested using chi2-test with 95% confident intervals. Our results indicated a statistically significant association (p<0.05) between rs1219648, but not rs1042522, and risk of breast cancer. We also found that the combination of FGFR2 major genotype and TP53 hetero genotype had protective effects against breast cancer , while the hetero allele of FGFR2 in combination with the minor genotype of TP53 was associated with a high risk. This study revealed an important crosstalk between two polymorphisms in FGFR2 and TP53 in development of breast cancer. These candidates risk variants should be further evaluated in studies with a larger sample size.