MilliporeSigma
  • Vinylic MIDA Boronates: New Building Blocks for the Synthesis of Aza-Heterocycles.
Vinylic MIDA Boronates: New Building Blocks for the Synthesis of Aza-Heterocycles.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-03-27)
Sabin Llona-Minguez, Matthieu Desroses, Artin Ghassemian, Sylvain A Jacques, Lars Eriksson, Rebecka Isacksson, Tobias Koolmeister, Pål Stenmark, Martin Scobie, Thomas Helleday
ABSTRACT
A two-step synthesis of structurally diverse pyrrole-containing bicyclic systems is reported. ortho-Nitro-haloarenes coupled with vinylic N-methyliminodiacetic acid (MIDA) boronates generate ortho-vinyl-nitroarenes, which undergo a "metal-free" nitrene insertion, resulting in a new pyrrole ring. This novel synthetic approach has a wide substrate tolerance and it is applicable in the preparation of more complex "drug-like" molecules. Interestingly, an ortho-nitro-allylarene derivative furnished a cyclic β-aminophosphonate motif.
Sigma-Aldrich
Vinylboronic acid MIDA ester, 97%