MilliporeSigma
  • Trimeric structure of PRL-1 phosphatase reveals an active enzyme conformation and regulation mechanisms.
Trimeric structure of PRL-1 phosphatase reveals an active enzyme conformation and regulation mechanisms.
Journal of molecular biology (2004-12-02)
Dae Gwin Jeong, Seung Jun Kim, Jae Hoon Kim, Jeong Hee Son, Mi Rim Park, Sang Myoun Lim, Tae-Sung Yoon, Seong Eon Ryu
ABSTRACT
The PRL phosphatases, which constitute a subfamily of the protein tyrosine phosphatases (PTPs), are implicated in oncogenic and metastatic processes. Here, we report the crystal structure of human PRL-1 determined at 2.7A resolution. The crystal structure reveals the shallow active-site pocket with highly hydrophobic character. A structural comparison with the previously determined NMR structure of PRL-3 exhibits significant differences in the active-site region. In the PRL-1 structure, a sulfate ion is bound to the active-site, providing stabilizing interactions to maintain the canonically found active conformation of PTPs, whereas the NMR structure exhibits an open conformation of the active-site. We also found that PRL-1 forms a trimer in the crystal and the trimer exists in the membrane fraction of cells, suggesting the possible biological regulation of PRL-1 activity by oligomerization. The detailed structural information on the active enzyme conformation and regulation of PRL-1 provides the structural basis for the development of potential inhibitors of PRL enzymes.