MilliporeSigma
  • C-Fos expression is a molecular predictor of progression and survival in epithelial ovarian carcinoma.
C-Fos expression is a molecular predictor of progression and survival in epithelial ovarian carcinoma.
British journal of cancer (2008-10-16)
S Mahner, C Baasch, J Schwarz, S Hein, L Wölber, F Jänicke, K Milde-Langosch
ABSTRACT
Members of the Fos protein family dimerise with Jun proteins to form the AP-1 transcription factor complex. They have a central function in proliferation and differentiation of normal tissue as well as in oncogenic transformation and tumour progression. We analysed the expression of c-Fos, FosB, Fra-1 and Fra-2 to investigate the function of Fos transcription factors in ovarian cancer. A total of 101 patients were included in the study. Expression of Fos proteins was determined by western blot analysis, quantified by densitometry and verified by immunohistochemistry. Reduced c-Fos expression was independently associated with unfavourable progression-free survival (20.6, 31.6 and 51.2 months for patients with low, moderate and high c-Fos expression; P=0.003) as well as overall survival (23.8, 46.0 and 55.5 months for low, moderate and high c-Fos levels; P=0.003). No correlations were observed for FosB, Fra-1 and Fra-2. We conclude that loss of c-Fos expression is associated with tumour progression in ovarian carcinoma and that c-Fos may be a prognostic factor. These results are in contrast to the classic concept of c-Fos as an oncogene, but are supported by the recently discovered tumour-suppressing and proapoptotic function of c-Fos in various cancer types.