MilliporeSigma
  • HIV-1 Tat protein induces glial cell autophagy through enhancement of BAG3 protein levels.
HIV-1 Tat protein induces glial cell autophagy through enhancement of BAG3 protein levels.
Cell cycle (Georgetown, Tex.) (2014-12-09)
Anna Paola Bruno, Francesca Isabella De Simone, Vittoria Iorio, Margot De Marco, Kamel Khalili, Ilker Kudret Sariyer, Mario Capunzo, Stefania Lucia Nori, Alessandra Rosati
ABSTRACT
BAG3 protein has been described as an anti-apoptotic and pro-autophagic factor in several neoplastic and normal cells. We previously demonstrated that BAG3 expression is elevated upon HIV-1 infection of glial and T lymphocyte cells. Among HIV-1 proteins, Tat is highly involved in regulating host cell response to viral infection. Therefore, we investigated the possible role of Tat protein in modulating BAG3 protein levels and the autophagic process itself. In this report, we show that transfection with Tat raises BAG3 levels in glioblastoma cells. Moreover, BAG3 silencing results in highly reducing Tat- induced levels of LC3-II and increasing the appearance of sub G0/G1 apoptotic cells, in keeping with the reported role of BAG3 in modulating the autophagy/apoptosis balance. These results demonstrate for the first time that Tat protein is able to stimulate autophagy through increasing BAG3 levels in human glial cells.