MilliporeSigma
  • Association of GGCX gene polymorphism with warfarin dose in atrial fibrillation population in Xinjiang.
Association of GGCX gene polymorphism with warfarin dose in atrial fibrillation population in Xinjiang.
Lipids in health and disease (2013-10-24)
Xiayizha Kamali, Muhuyati Wulasihan, Yu-Chun Yang, Wu-Hong Lu, Zhi-Qiang Liu, Peng-Yi He
ABSTRACT
To study the effects of γ-glutamyl carboxylase (GGCX) rs2592551 polymorphism on warfarin dose in atrial fibrillation patients in Xinjiang region. Polymerase chain reaction - restriction fragment length polymorphism and direct sequencing methods were used to detect the rs2592551 genotype in 269 atrial fibrillation patients with warfarin administration. The effects of different genotypes on warfarin dose were statistically analyzed. The rs2592551 polymorphism detection results were 136 cases of wild-type homozygous CC genotype (50.56%), 115 cases of heterozygous CT genotype (42.75%), 18 cases of homozygous TT genotype (6.69%). The allele frequency C was 71.93%, T was 28.07%. The stable warfarin dose average was 2.86 ± 0.61 mg/d in patients with CC genotype, 3.59 ± 0.93 mg/d in patients with CT genotype and 4.06 ± 0.88 mg/d in patients with TT genotype. The warfarin dose in different genotypes were compared, there was statistically significant difference between CC and TT, CC and CT (P <0. 05), but the TT and CT showed no significant difference (P > 0.05). In atrial fibrillation population in Xinjiang, patients with CT and TT genotypes in GGCX gene rs259251 loci required for significantly higher warfarin dose than those with CC genotype. Therefore, rs2592551 polymorphism may one of the factors affecting the warfarin dose in patients with atrial fibrillation.