MilliporeSigma
  • Identification of novel oligodendroglioma-associated candidate tumor suppressor genes in 1p36 and 19q13 using microarray-based expression profiling.
Identification of novel oligodendroglioma-associated candidate tumor suppressor genes in 1p36 and 19q13 using microarray-based expression profiling.
International journal of cancer (2006-03-22)
Bjoern Tews, Joerg Felsberg, Christian Hartmann, Annegret Kunitz, Meinhard Hahn, Grischa Toedt, Kai Neben, Lars Hummerich, Andreas von Deimling, Guido Reifenberger, Peter Lichter
ABSTRACT
Loss of heterozygosity (LOH) on chromosomal arms 1p and 19q is the most common genetic alteration in oligodendroglial tumors and associated with response to radio- and chemotherapy as well as favorable prognosis. Using microsatellite analysis, we previously identified the chromosomal regions 1p36.22-p36.31 and 19q13.3, as candidate tumor suppressor gene regions being commonly deleted in these tumors. To identify genes within these regions that are downregulated in oligodendroglial tumors with LOH 1p/19q, we performed cDNA microarray-based RNA expression profiling of 35 gliomas with known allelic status on 1p and 19q, including 7 oligodendrogliomas and 8 diffuse astrocytomas of World Health Organization (WHO) grade II, as well as 14 anaplastic oligodendrogliomas and 6 anaplastic oligoastrocytomas of WHO grade III. The microarrays used for expression profiling carried approximately 7,000 gene-specific cDNAs, with complete coverage of the genes located in 1p36.13-p36.31 and 19q13.2-q13.33. Microarray analysis identified 8 genes from these regions (MGC4399, SRM, ICMT, RPL18, FTL, ZIN, FLJ10781 and DBP), which all showed significantly lower expression in 1p/19q-deleted gliomas when compared to gliomas without 1p/19q losses. Quantitative real-time reverse transcription-PCR analyses were performed for the MGC4399, ICMT and RPL18 genes and confirmed the microarray findings. In addition, we found that the cytosolic phospholipase A2 (PLA2G4C) gene at 19q13.3 demonstrated significantly lower expression in anaplastic oligodendrogliomas (WHO grade III) when compared to well-differentiated oligodendrogliomas (WHO grade II). Taken together, our study provides a set of interesting novel candidate genes that may play important roles in the pathogenesis of oligodendroglial tumors.