MilliporeSigma
  • Stable isotopic profiling of intermediary metabolic flux in developing and adult stage Caenorhabditis elegans.
Stable isotopic profiling of intermediary metabolic flux in developing and adult stage Caenorhabditis elegans.
Journal of visualized experiments : JoVE (2011-03-16)
Marni J Falk, Meera Rao, Julian Ostrovsky, Evgueni Daikhin, Ilana Nissim, Marc Yudkoff
ABSTRACT
Stable isotopic profiling has long permitted sensitive investigations of the metabolic consequences of genetic mutations and/or pharmacologic therapies in cellular and mammalian models. Here, we describe detailed methods to perform stable isotopic profiling of intermediary metabolism and metabolic flux in the nematode, Caenorhabditis elegans. Methods are described for profiling whole worm free amino acids, labeled carbon dioxide, labeled organic acids, and labeled amino acids in animals exposed to stable isotopes either from early development on nematode growth media agar plates or beginning as young adults while exposed to various pharmacologic treatments in liquid culture. Free amino acids are quantified by high performance liquid chromatography (HPLC) in whole worm aliquots extracted in 4% perchloric acid. Universally labeled (13)C-glucose or 1,6-(13)C(2)-glucose is utilized as the stable isotopic precursor whose labeled carbon is traced by mass spectrometry in carbon dioxide (both atmospheric and dissolved) as well as in metabolites indicative of flux through glycolysis, pyruvate metabolism, and the tricarboxylic acid cycle. Representative results are included to demonstrate effects of isotope exposure time, various bacterial clearing protocols, and alternative worm disruption methods in wild-type nematodes, as well as the relative extent of isotopic incorporation in mitochondrial complex III mutant worms (isp-1(qm150)) relative to wild-type worms. Application of stable isotopic profiling in living nematodes provides a novel capacity to investigate at the whole animal level real-time metabolic alterations that are caused by individual genetic disorders and/or pharmacologic therapies.
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes