MilliporeSigma
  • Effects of low incubation temperatures on the bactericidal activity of anti-tuberculosis drugs.
Effects of low incubation temperatures on the bactericidal activity of anti-tuberculosis drugs.
The Journal of antimicrobial chemotherapy (2010-11-13)
David Coleman, Simon J Waddell, Denis A Mitchison
ABSTRACT
to explore the effect of low incubation temperatures and the consequent slowing of bacterial metabolism on the bactericidal action of anti-tuberculosis drugs against Mycobacterium tuberculosis. counting of surviving bacteria during exposure of static cultures to 1 mg/L isoniazid, 2 mg/L rifampicin, 0.5 or 2 mg/L TMC207 and 40 or 160 mg/L pyrazinamide, usually for periods of 21 days at temperatures of 37, 25, 22, 19, 16 or 8°C. the bactericidal activities of isoniazid and rifampicin were progressively reduced at 25 and 22°C, and were minimal at lower temperatures. TMC207 was immediately bactericidal at 37°C, in contrast to the early static phase reported with log phase cultures, and showed less change in activity as incubation temperatures were reduced than did rifampicin or isoniazid. Pyrazinamide was more bactericidal when incubation temperatures were reduced below 37°C and when the static seed cultures were most dormant. these results can be explained by the surmise that at low temperatures bacterial energy is at a low level with only just sufficient ATP to maintain homeostasis, making the bacteria more susceptible to the blocking of ATP synthesis by TMC207. Insufficient ATP at low temperature would also hinder the export of pyrazinoic acid, the toxic product of the pro-drug pyrazinamide, from the mycobacterial cell by an inefficient efflux pump that requires energy.
Sigma-Aldrich
Carbendazim, 97%