MilliporeSigma
  • Transforming Suzuki-Miyaura cross-couplings of MIDA boronates into a green technology: no organic solvents.
Transforming Suzuki-Miyaura cross-couplings of MIDA boronates into a green technology: no organic solvents.
Journal of the American Chemical Society (2013-11-15)
Nicholas A Isley, Fabrice Gallou, Bruce H Lipshutz
ABSTRACT
New technology has been developed that enables Suzuki-Miyaura couplings involving widely utilized MIDA boronates to be run in water as the only medium, mainly at room temperature. The protocol is such that no organic solvent is involved at any stage; from the reaction through to product isolation. Hence, using the E factor scale as a measure of greenness, the values for these cross-couplings approach zero.