MilliporeSigma
  • Living vs. deceased donor liver transplantation provides comparable recovery of renal function in patients with hepatorenal syndrome: a matched case-control study.
Living vs. deceased donor liver transplantation provides comparable recovery of renal function in patients with hepatorenal syndrome: a matched case-control study.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2014-10-04)
N Goldaracena, M Marquez, N Selzner, V N Spetzler, M S Cattral, P D Greig, L Lilly, I D McGilvray, G A Levy, A Ghanekar, E L Renner, D R Grant, M Selzner
ABSTRACT
Outcomes of living versus deceased donor liver transplantation in patients with chronic liver disease and hepatorenal syndrome (HRS) was compared using a matched pair study design. Thirty patients with HRS receiving a live donor liver transplantation (LDLT) and 90 HRS patients receiving a full graft deceased donor liver transplantation (DDLT) were compared. LDLT versus DDLT of patients with HRS was associated with decreased peak aspartate aminotransferase levels (339 ± 214 vs. 935 ± 1253 U/L; p = 0.0001), and similar 7-day bilirubin (8.42 ± 7.89 vs. 6.95 ± 7.13 mg/dL; p = 0.35), and international normalized ratio levels (1.93 ± 0.62 vs. 1.78 ± 0.78; p = 0.314). LDLT vs. DDLT had a decreased intensive care unit (2 [1-39] vs. 4 [0-93] days; p = 0.004), and hospital stay (17 [4-313] vs. 26 [0-126] days; p = 0.016) and a similar incidence of overall postoperative complications (20% vs. 27%; p = 0.62). No difference was detected between LDLT and DDLT patients regarding graft survival at 1 (80% vs. 82%), at 3 (69% vs. 76%) and 5 years (65% vs. 76%) (p = 0.63), as well as patient survival at 1 (83% vs. 82%), 3 (72% vs. 77%) and 5 years (72% vs. 77%) (p = 0.93). The incidence of chronic kidney disease post-LT (10% vs. 6%; p = 0.4) was similar between both groups. LDLT results in identical long-term outcome when compared with DDLT in patients with HRS.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder