MilliporeSigma
  • Quantifying the effects of inactin vs Isoflurane anesthesia on gastrointestinal motility in rats using dynamic magnetic resonance imaging and spatio-temporal maps.
Quantifying the effects of inactin vs Isoflurane anesthesia on gastrointestinal motility in rats using dynamic magnetic resonance imaging and spatio-temporal maps.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2014-09-27)
A C Ailiani, T Neuberger, J G Brasseur, G Banco, Y Wang, N B Smith, A G Webb
ABSTRACT
Anesthetics are commonly applied in animal studies of gastrointestinal (GI) function. Different anesthetics alter smooth-muscle motility in different ways. The aim of this study is to quantify and compare non-invasively with magnetic resonance imaging (MRI) the motility patterns of the rat gut when anesthetized with inactin vs isoflurane anesthetics in the fed state. Rats were given an oral gavage of MRI contrast agent for improved visualization of the GI tract. Two-dimensional images through the jejunum of the pre- and postanesthetized rat in the fed state were acquired every 168 ms. Image registration, segmentation, and postprocessing algorithms were applied to produce spatio-temporal maps that were used to quantify peristaltic and segmental motions in the jejunum region interspersed between periods of inactivity. There were significantly longer periods of inactivity in the rats treated with isoflurane than in those treated with inactin (179.9 ± 22.4 s vs 17.7 ± 10.3 s). The speed of propagation and wavelength of peristalsis, and the frequency and speed of pattern switching of segmental motility, were higher (p < 0.05) in rats treated with inactin. Isoflurane and inactin anesthetics produce significantly different motility behavior with the rat's GI tract in the fed state. Isoflurane anesthetic, results in a reduced frequency of occurrence of motility periods and an overall reduced level of motility in comparison with inactin.
Sigma-Aldrich
Inactin® hydrate, ≥98% (HPLC)