MilliporeSigma
  • Urinary p-cresol is elevated in young French children with autism spectrum disorder: a replication study.
Urinary p-cresol is elevated in young French children with autism spectrum disorder: a replication study.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2014-07-11)
Stefano Gabriele, Roberto Sacco, Sonia Cerullo, Cristina Neri, Andrea Urbani, Gabriele Tripi, Joëlle Malvy, Catherine Barthelemy, Frédérique Bonnet-Brihault, Antonio M Persico
ABSTRACT
The aromatic compound p-cresol (4-methylphenol) has been found elevated in the urines of Italian autistic children up to 8 years of age. The present study aims at replicating these initial findings in an ethnically distinct sample and at extending them by measuring also the three components of urinary p-cresol, namely p-cresylsulfate, p-cresylglucuronate and free p-cresol. Total urinary p-cresol, p-cresylsulfate and p-cresylglucuronate were significantly elevated in 33 French autism spectrum disorder (ASD) cases compared with 33 sex- and age-matched controls (p < 0.05). This increase was limited to ASD children aged ≤8 years (p < 0.01), and not older (p = 0.17). Urinary levels of p-cresol and p-cresylsulfate were associated with stereotypic, compulsive/repetitive behaviors (p < 0.05), although not with overall autism severity. These results confirm the elevation of urinary p-cresol in a sizable set of small autistic children and spur interest into biomarker roles for p-cresol and p-cresylsulfate in autism.
Sigma-Aldrich
2-tert-Butylphenol, 99%
Pricing and availability is not currently available.