MilliporeSigma
  • NADPH oxidase DUOX1 promotes long-term persistence of oxidative stress after an exposure to irradiation.
NADPH oxidase DUOX1 promotes long-term persistence of oxidative stress after an exposure to irradiation.
Proceedings of the National Academy of Sciences of the United States of America (2015-04-08)
Rabii Ameziane-El-Hassani, Monique Talbot, Maria Carolina de Souza Dos Santos, Abir Al Ghuzlan, Dana Hartl, Jean-Michel Bidart, Xavier De Deken, Françoise Miot, Ibrahima Diallo, Florent de Vathaire, Martin Schlumberger, Corinne Dupuy
ABSTRACT
Ionizing radiation (IR) causes not only acute tissue damage, but also late effects in several cell generations after the initial exposure. The thyroid gland is one of the most sensitive organs to the carcinogenic effects of IR, and we have recently highlighted that an oxidative stress is responsible for the chromosomal rearrangements found in radio-induced papillary thyroid carcinoma. Using both a human thyroid cell line and primary thyrocytes, we investigated the mechanism by which IR induces the generation of reactive oxygen species (ROS) several days after irradiation. We focused on NADPH oxidases, which are specialized ROS-generating enzymes known as NOX/DUOX. Our results show that IR induces delayed NADPH oxidase DUOX1-dependent H2O2 production in a dose-dependent manner, which is sustained for several days. We report that p38 MAPK, activated after IR, increased DUOX1 via IL-13 expression, leading to persistent DNA damage and growth arrest. Pretreatment of cells with catalase, a scavenger of H2O2, or DUOX1 down-regulation by siRNA abrogated IR-induced DNA damage. Analysis of human thyroid tissues showed that DUOX1 is elevated not only in human radio-induced thyroid tumors, but also in sporadic thyroid tumors. Taken together, our data reveal a key role of DUOX1-dependent H2O2 production in long-term persistent radio-induced DNA damage. Our data also show that DUOX1-dependent H2O2 production, which induces DNA double-strand breaks, can cause genomic instability and promote the generation of neoplastic cells through its mutagenic effect.
Sigma-Aldrich
MISSION® esiRNA, targeting human IL13
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Duox2
Sigma-Aldrich
MISSION® esiRNA, targeting human DUOX1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Duox1