MilliporeSigma
  • Identification and phytotoxicity of a new glucosinolate breakdown product from Meadowfoam (Limnanthes alba) seed meal.
Identification and phytotoxicity of a new glucosinolate breakdown product from Meadowfoam (Limnanthes alba) seed meal.
Journal of agricultural and food chemistry (2014-07-08)
Suphannika Intanon, Ralph L Reed, Jan F Stevens, Andrew G Hulting, Carol A Mallory-Smith
ABSTRACT
Meadowfoam (Limnanthes alba Hartw. ex Benth.) is an oilseed crop grown in the Willamette Valley of Oregon. Meadowfoam seed meal (MSM), a byproduct after oil extraction, contains 2-4% glucosinolate (glucolimnanthin). Activated MSM, produced by adding freshly ground myrosinase-active meadowfoam seeds to MSM, facilitates myrosinase-mediated formation of glucosinolate-derived degradation products with herbicidal activity. In the activated MSM, glucolimnanthin was converted into 3-methoxybenzyl isothiocyanate ("isothiocyanate") within 24 h and was degraded by day three. 3-Methoxyphenylacetonitrile ("nitrile") persisted for at least 6 days. Methoxyphenylacetic acid (MPAA), a previously unknown metabolite of glucolimnanthin, appeared at day three. Its identity was confirmed by LC-UV and high resolution LC-MS/MS comparisons with a standard of MPAA. Isothiocyanate inhibited lettuce germination 8.5- and 14.4-fold more effectively than MPAA and nitrile, respectively. Activated MSM inhibited lettuce germination by 58% and growth by 72% compared with the control. Results of the study suggest that MSM has potential uses as a pre-emergence bioherbicide.
Sigma-Aldrich
3-Methoxyphenylacetonitrile, 99%
Pricing and availability is not currently available.