MilliporeSigma
  • Healthcare-associated and nosocomial bacterial infections in cirrhosis: predictors and impact on outcome.
Healthcare-associated and nosocomial bacterial infections in cirrhosis: predictors and impact on outcome.
Liver international : official journal of the International Association for the Study of the Liver (2014-07-22)
Konstantina Sargenti, Hanne Prytz, Anna Strand, Emma Nilsson, Evangelos Kalaitzakis
ABSTRACT
Population-based data on the occurrence of healthcare-associated (HCA) and hospital-acquired (HA) bacterial infections in cirrhosis, their predictors, and their impact on outcome are limited. All patients with incident cirrhosis in 2001-2010 residing in an area of 600,000 inhabitants were retrospectively identified. All serious bacterial infections (resulting in or occurring during an inpatient hospital episode) during this period were registered. Acquisition type, site of infection, occurrence of infection-related acute-on-chronic liver failure (ACLF), acute kidney injury (AKI) and bacterial resistance were analysed. Patients were followed longitudinally until death, transplant or end of 2011. A total of 398 serious infections occurred in 241/633 (38%) patients. Forty-seven per cent were HCA and 21% HA. Proton pump inhibitor (PPI) use was more common in HA (80%) vs. HCA (64%) vs. community-acquired (44%) infections (P < 0.001). In regression analysis, decompensated status, use of antibiotics and PPIs at infection diagnosis were independent predictors of HCA/HA infections (P < 0.05). After adjustment for confounders, HCA/HA infections were significantly related to infection-related ACLF (P < 0.05), but not severe sepsis, AKI or infection-related mortality (P > 0.05). Antibiotic-resistant infections were more frequent among HA (17%) than HCA (6%) or community-acquired (8%) infections (P < 0.05). Antibiotic-resistant HCA/HA infections were independently related to severe sepsis (P < 0.05). In a population-based cirrhotic cohort, two-thirds of serious bacterial infections were HCA or HA. Decompensated liver disease, antibiotics and PPIs were predictors of serious HCA/HA infections, which were associated with the development of ACLF. Antibiotic resistance was frequent, especially in HA infections, and contributed to risk of severe sepsis.
Sigma-Aldrich
Hexachloro-2-propanone, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Hexachloro-2-propanone, 99%