MilliporeSigma
  • Nanotopographical induction of osteogenesis through adhesion, bone morphogenic protein cosignaling, and regulation of microRNAs.
Nanotopographical induction of osteogenesis through adhesion, bone morphogenic protein cosignaling, and regulation of microRNAs.
ACS nano (2014-09-18)
Jingli Yang, Laura E McNamara, Nikolaj Gadegaard, Enateri V Alakpa, Karl V Burgess, R M Dominic Meek, Matthew J Dalby
ABSTRACT
It is emerging that nanotopographical information can be used to induce osteogenesis from mesenchymal stromal cells from the bone marrow, and it is hoped that this nanoscale bioactivity can be utilized to engineer next generation implants. However, the osteogenic mechanism of surfaces is currently poorly understood. In this report, we investigate mechanism and implicate bone morphogenic protein (BMP) in up-regulation of RUNX2 and show that RUNX2 and its regulatory miRNAs are BMP sensitive. Our data demonstrate that osteogenic nanotopography promotes colocalization of integrins and BMP2 receptors in order to enhance osteogenic activity and that vitronectin is important in this interface. This provides insight that topographical regulation of adhesion can have effects on signaling cascades outside of cytoskeletal signaling and that adhesions can have roles in augmenting BMP signaling.
Sigma-Aldrich
Anti-RUNX2 Antibody, clone AS110, clone AS110, from mouse
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography