MilliporeSigma
Periostin induces pancreatic regeneration.
Endocrinology (2014-12-09)
Johnathan K Smid, Sharlene Faulkes, Michael A Rudnicki
ABSTRACT
We found that the secreted protein periostin (Postn) is highly induced after partial pancreatectomy in regenerating areas containing mesenchymal stroma and tubular complexes. Importantly, after partial pancreatectomy, Postn-deficient mice exhibit impaired mesenchymal formation and reduced regeneration specifically within the pancreatic β-cell compartment. Furthermore, Postn-deficient mice demonstrate an increased sensitivity to streptozotocin. Notably, injection of Postn directly into the pancreas stimulated proliferation of vimentin-expressing cells within 24 hours, and by 3 days, a mesenchymal stroma was present containing proliferating duct-like cells expressing the progenitor markers Ngn3 and Pdx1. Intraperitoneal injection of Postn resulted in increased numbers of islets and long-term glucoregulatory benefits with no adverse effects found in other tissues. Delivery of Postn throughout the pancreas via the common bile duct resulted in increased numbers of small insulin-expressing clusters and a significant improvement in glucose tolerance. Therefore, Postn is novel molecule capable of potentiating pancreatic β-cell regeneration.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder