MilliporeSigma
IKK-ε coordinates invasion and metastasis of ovarian cancer.
Cancer research (2012-09-04)
Sarah Hsu, Marianne Kim, Lidia Hernandez, Valentina Grajales, Anne Noonan, Miriam Anver, Ben Davidson, Christina M Annunziata
ABSTRACT
Inhibitor of IκB kinases (IKK) are key regulators of NF-κB signaling. Three IKK isoforms-α, β, and ε-have been linked to oncogenesis, yet the precise components of NF-κB signaling in ovarian cancer have not yet been dissected. We surveyed 120 ovarian cancer specimens for IKK-ε expression. Notably, cytoplasmic expression was elevated in metastatic lesions relative to primary tumors (P = 0.03). Therefore, we hypothesized that IKK-ε drives ovarian cancer metastasis. IKK-ε was identified previously as a breast cancer oncogene and was associated with poor clinical outcome in ovarian cancer. We now define an ovarian cancer-specific IKK-ε-regulated gene expression signature using stably expressed short hairpin RNA targeting IKK-ε. Pathway analysis of the signature indicated that IKK-ε regulates expression of genes involved in cell motility and inflammation. We further showed that IKK-ε depletion in metastatic ovarian cancer cell lines decreased growth, adhesion, and invasion. Consistently, human xenografts depleted of IKK-ε in mice showed decreased aggressiveness, whereas overexpression of IKK-ε in a less invasive ovarian cancer cell line increased metastasis in vivo. Taken together, these data provide evidence that IKK-ε is a key coordinator of invasion and metastasis programs in ovarian cancer. Inhibition of IKK-ε signaling thus emerges as a viable therapeutic strategy in women whose ovarian cancer shows aberrant activation of this pathway.