MilliporeSigma
  • Studies of P(L/D)LA 96/4 non-woven scaffolds and fibres; properties, wettability and cell spreading before and after intrusive treatment methods.
Studies of P(L/D)LA 96/4 non-woven scaffolds and fibres; properties, wettability and cell spreading before and after intrusive treatment methods.
Journal of materials science. Materials in medicine (2007-02-06)
Ville Ellä, Manuela E Gomes, Rui L Reis, Pertti Törmälä, Minna Kellomäki
ABSTRACT
Poly(L/D)lactide 96/4 fibres with diameters of 50 and 80 microm were produced. The smaller diameter fibres were carded and needle punched to form a non-woven mat. Fibres and non-woven mats were hydrolysed for a period of 20 weeks. Fibres and pressed non-woven discs were treated with low-temperature oxygen plasma and alkaline KOH hydrolysis and ethanol washing was used as a reference treatment. The non-wovens lost 50% of their tear strength after 8 weeks in vitro while the fibres still retained 65% tensile strength after 20 weeks. Hydrolysation time in KOH, treatment time and power settings of the oxygen plasma were all directly proportional to the mechanical properties of the fibres. Increasing time (and power) resulted in lower tensile properties. Rapid wetting of the scaffolds was achieved by oxygen plasma, KOH hydrolysation and ethanol washing. Cell culturing using fibroblast cell line was carried out for the treated and non-treated non-woven scaffolds. In terms of adhesion and the spreading of the cells into the scaffold, best results after 3-day culturing were obtained for the oxygen plasma treated scaffolds.
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Pricing and availability is not currently available.