MilliporeSigma
  • Activation of the P2Y1 receptor induces apoptosis and inhibits proliferation of prostate cancer cells.
Activation of the P2Y1 receptor induces apoptosis and inhibits proliferation of prostate cancer cells.
Biochemical pharmacology (2011-06-03)
Qiang Wei, Stefano Costanzi, Qiu-Zhen Liu, Zhan-Guo Gao, Kenneth A Jacobson
ABSTRACT
G protein-coupled receptors, the largest cell surface receptor family, have emerged as critical players in cell death and survival. High gene expression level of the G(q)-coupled P2Y(1) nucleotide receptor in PC-3 prostate cancer cells was demonstrated using real-time quantitative PCR and confirmed by Western blotting and confocal laser scanning microscopy. A selective P2Y(1) receptor agonist, the ADP analogue MRS2365, concentration-dependently induced intracellular calcium mobilization (EC(50) 5.28nM), which was diminished by P2Y(1) receptor-selective antagonist MRS2500. P2Y(1) receptor activation by MRS2365 induced apoptosis in assays of Caspase-3, LDH release, and annexin-V staining. The pro-apoptotic effect of MRS2365 was blocked by MRS2500, P2Y(1) siRNA, and an inhibitor of the MAP kinase pathway PD98059. MRS2365 significantly inhibited the proliferation of PC-3 cells, examined using a MTT assay. Thus, activation of the P2Y(1) receptor induced cell death and inhibited growth of human prostatic carcinoma PC-3 cells. Activation of the P2Y(1) receptor should be a novel and promising therapeutic strategy for prostate cancer.