MilliporeSigma
  • Primary structure and functional activity of a phosphatidylinositol-glycan-specific phospholipase D.
Primary structure and functional activity of a phosphatidylinositol-glycan-specific phospholipase D.
Science (New York, N.Y.) (1991-04-19)
B J Scallon, W J Fung, T C Tsang, S Li, H Kado-Fong, K S Huang, J P Kochan
ABSTRACT
A phosphatidylinositol-glycan-specific phospholipase D (PI-G PLD) that specifically hydrolyzes the inositol phosphate linkage in proteins anchored by phosphatidylinositol-glycans (PI-Gs) has recently been purified from human and bovine sera. The primary structure of bovine PI-G PLD has now been determined and the functional activity of the enzyme has been studied. Expression of PI-G PLD complementary DNA in COS cells produced a protein that specifically hydrolyzed the inositol phosphate linkage of the PI-G anchor. Cotransfection of PI-G PLD with a PI-G-anchored protein resulted in the secretion of the PI-G-anchored protein. The results suggest that the expression of PI-G PLD may influence the expression and location of PI-G-anchored proteins.