MilliporeSigma
  • Identification of the human cytochrome P450, P450RAI-2, which is predominantly expressed in the adult cerebellum and is responsible for all-trans-retinoic acid metabolism.
Identification of the human cytochrome P450, P450RAI-2, which is predominantly expressed in the adult cerebellum and is responsible for all-trans-retinoic acid metabolism.
Proceedings of the National Academy of Sciences of the United States of America (2000-05-24)
J A White, H Ramshaw, M Taimi, W Stangle, A Zhang, S Everingham, S Creighton, S P Tam, G Jones, M Petkovich
ABSTRACT
Retinoids, particularly all-trans-retinoic acid (RA), are potent regulators of cell differentiation, cell proliferation, and apoptosis. The role of all-trans-RA during development and in the maintenance of adult tissues has been well established. The control of all-trans-RA levels in cells and tissues is regulated by the balance between its biosynthesis and its catabolism to inactive metabolites. The cytochrome P450 enzyme P450RAI (herein renamed P450RAI-1) is partially responsible for this inactivation of all-trans-RA. In this report, we describe the identification, molecular cloning, and characterization of a second related enzyme, P450RAI-2, which is also involved in the specific inactivation of all-trans-RA. Transiently transfected P450RAI-2 can convert all-trans-RA to more polar metabolites including 4-oxo-, 4-OH-, and 18-OH-all-trans-RA. Competition experiments with other retinoids suggest that all-trans-RA is the preferred substrate. The high level of expression of P450RAI-2, particularly in the cerebellum and pons of human adult brain, suggests a unique role for this enzyme in the protection of specific tissues from exposure to retinoids.