MilliporeSigma
  • CSF1R mutations in hereditary diffuse leukoencephalopathy with spheroids are loss of function.
CSF1R mutations in hereditary diffuse leukoencephalopathy with spheroids are loss of function.
Scientific reports (2013-10-23)
Clare Pridans, Kristin A Sauter, Kristin Baer, Holger Kissel, David A Hume
ABSTRACT
Hereditary diffuse leukoencephalopathy with spheroids (HDLS) in humans is a rare autosomal dominant disease characterized by giant neuroaxonal swellings (spheroids) within the CNS white matter. Symptoms are variable and can include personality and behavioural changes. Patients with this disease have mutations in the protein kinase domain of the colony-stimulating factor 1 receptor (CSF1R) which is a tyrosine kinase receptor essential for microglia development. We investigated the effects of these mutations on Csf1r signalling using a factor dependent cell line. Corresponding mutant forms of murine Csf1r were expressed on the cell surface at normal levels, and bound CSF1, but were not able to sustain cell proliferation. Since Csf1r signaling requires receptor dimerization initiated by CSF1 binding, the data suggest a mechanism for phenotypic dominance of the mutant allele in HDLS.
Sigma-Aldrich
Human CSF1R / Macrophage Colony-stimulating Factor 1 Receptor ELISA Kit
Sigma-Aldrich
FMS (539-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution