MilliporeSigma
  • Molecular and genetic analysis of the transferred DNA regions of the root-inducing plasmid of Agrobacterium rhizogenes.
Molecular and genetic analysis of the transferred DNA regions of the root-inducing plasmid of Agrobacterium rhizogenes.
Journal of bacteriology (1985-10-01)
F F White, B H Taylor, G A Huffman, M P Gordon, E W Nester
ABSTRACT
The T-DNA regions of the root-inducing (Ri) plasmid pRiA4b of Agrobacterium rhizogenes were characterized. Two regions, designated TL-DNA and TR-DNA, were found to be integrated and stably maintained in the plant genome. The TL-DNA spanned a 15- to 20-kilobase region of pRiA4b and was separated from the TR-DNA region by at least 15 kilobases of nonintegrated plasmid DNA. The TR-DNA region also spanned a 15- to 20-kilobase region of pRiA4b and included a region of homology to the tms morphogenic loci of the tumor-inducing (Ti) plasmid of Agrobacterium tumefaciens. Eighteen deletions and 95 transposon insertions were generated in the T-DNA regions and tested for alterations in virulence. Insertions into four loci in the TL-DNA affected the morphology of root formation of Kalanchoë diagremontiana leaves and stems, but had no visible effects on other host plants. Insertions into two loci (tms-1 and tms-2) in the TR-DNA eliminated virulence symptoms on all plants tested, with the exception of K. diagremontiana stems, where sparse root formation occurred. Complementation experiments with Ri and Ti plasmid T-DNA mutations indicate that the tms genes of the two plasmids serve similar functions and suggest a functional relationship between one or more genes of the TL-DNA and the cytokinin synthesis locus tmr of the Ti plasmid.
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Pricing and availability is not currently available.