- Enzymatic synthesis of trehalose esters having lipophilicity.
Enzymatic synthesis of trehalose esters having lipophilicity.
To improve trehalose lipophilicity, trehalose was regioselectively esterified with vinyl fatty acid esters in dimethyl formamide by protease from Bacillus subtilis to give 6-O-lauroyltrehalose, 6-O-myristoyltrehalose, 6-O-palmitoyltrehalose, 6-O-stearoyltrehalose, 6-O-oleoyltrehalose and 6-O-linoleoyltrehalose. The influence of structural variation by changing fatty acid substitute was examined by measurement of the surface tension and biodegradability.