MilliporeSigma
  • The gene encoding the immunoregulatory signaling molecule CMRF-35A localized to human chromosome 17 in close proximity to other members of the CMRF-35 family.
The gene encoding the immunoregulatory signaling molecule CMRF-35A localized to human chromosome 17 in close proximity to other members of the CMRF-35 family.
Tissue antigens (2001-09-15)
G J Clark, B Cooper, S Fitzpatrick, B J Green, D N Hart
ABSTRACT
The immunoregulatory signaling (IRS) family includes several molecules, which play major roles in the regulation of the immune response. The CMRF-35A and CMRF-35H molecules are two new members of the IRS family of molecules, that are found on a wide variety of haemopoietic lineages. The extracellular functional interactions of these molecules is presently unknown, although CMRF-35H can initiate an inhibitory signal and is internalized when cross-linked. In this paper, we described the gene structure for the CMRF-35A gene and its localization to human chromosome 17. The gene consists of four exons spanning approximately 4.5 kb. Exon 1 encodes the 5' untranslated region and leader sequence, exon 2 encodes the immunoglobulin (Ig)-like domain, exon 3 encodes the membrane proximal region and exon 4 encodes the transmembrane region, the cytoplasmic tail and the 3' untranslated region. A region in the 5' flanking sequence of the CMRF-35A gene, that promoted expression of a reporter gene was identified. The genes for the CMRF-35A and CMRF-35H molecules are closely linked on chromosome 17. Similarity between the Ig-like exons and the preceding intron of the two genes suggests exon duplication was involved in their evolution. We also identified a further member of the CMRF-35 family, the CMRF-35J pseudogene. This gene appears to have arisen by gene duplication of the CMRF-35A gene. These three loci - the CMRF-35A, CMRF-35J and CMRF-35H genes-form a new complex of IRS genes on chromosome 17.