MilliporeSigma
Detection of zona pellucida proteins during human folliculogenesis.
Human reproduction (Oxford, England) (2007-11-24)
Debra A Gook, D H Edgar, J Borg, M Martic
ABSTRACT
The stage of folliculogenesis at which the human zona pellucida (ZP) is initiated and the cells responsible for the origin of the ZP continue to be controversial. This study characterizes the development of the ZP during human folliculogenesis using ovarian samples donated from patients requesting ovarian storage. Follicles (from n = 18 patients, 14-40 years old) within fresh tissue and following development in a xenograft system were stained, using immunohistochemical techniques, for the presence of the three human ZP proteins, ZP1, ZP2 and ZP3. Over 500 primordial follicles and >20 follicles at each developmental stage were examined. All three ZP proteins were detected within the oocyte of the primordial follicle. Presence of ZP1 and ZP3 was observed in the majority of primordial oocytes (93% and 95%, respectively), whereas ZP2 was detected in only 32% of these follicles. The three ZP proteins were detected in the cytoplasm of cuboidal granulosa cells and their distribution correlates with developmental stages throughout folliculogenesis. ZP proteins were detected in both the oocyte and the granulosa cells as early as the primordial follicle stage in the human. The detection of ZP proteins in the quiescent primordial follicle suggests that these proteins have been present since oogenesis.