MilliporeSigma
  • Protocadherin-10 is involved in angiogenesis and methylation correlated with multiple myeloma.
Protocadherin-10 is involved in angiogenesis and methylation correlated with multiple myeloma.
International journal of molecular medicine (2012-01-17)
Ying Li, Ze-song Yang, Jun-jun Song, Qiong Liu, Jian-bin Chen
ABSTRACT
Protocadherin-10 (PCDH10) which is located at 4q28.3, is a member of the cadherin superfamily of cell adhesion molecules. PCDH10 is broadly expressed in normal adult, but nearly undetectable in multiple myeloma (ΜΜ) tissues and cell lines. Its promoter methylation was detected in virtually all the silenced or downregulated cell lines. The silencing of PCDH10 could be reversed by pharmacological demethylation, indicating a methylation-mediated mechanism. In the current study, we investigated 44 patients (23 females, 21 males), 77.27% (34/44) of whom presented high methylation of PCDH10. We found no associations between promoter hypermethylation and gender or age at the time of initial diagnosis. We also examined the role of PCDH10 as a mediator of MM cell proliferation, cell cycle progression, and its involvement in angiogenesis. Our results demonstrate that the PCDH10 gene is a target for epigenetic silencing in MM and provide a link between the dysregulation of angiogenesis and DNA methylation.
Sigma-Aldrich
MISSION® esiRNA, targeting human PCDH10