MilliporeSigma
Trichloroethylene. I. An overview.
Journal of toxicology and environmental health (1977-01-01)
E M Waters, H B Gerstner, J E Huff
PMID403297
ABSTRACT
Trichloroethylene (TCE) has been an industrial chemical of some importance for the past 50 years. First synthesized by Fischer in 1864, TCE has enjoyed considerable industrial usage as a degreaser and limited medical use as an inhalation anesthetic and analgesic. This TCE overview provides a narrative survey of the reference literature. Highlights include history, nomenclature, physical and chemical properties, manufacture, analysis, uses, metabolism, toxicology, carcinogenic potential, exposure routes, recommended standards, and conclusions. Chemically, TCE is a colorless, highly volatile liquid of molecular formula C2HCl3. Autoxidation of the unstable compound yields acidic products. Stabilizers are added to retard decomposition. TCE's multitude of industrial uses center around its highly effective fat-solvent properties. Metabolically, TCE is transformed in the liver to trichloroacetic acid, trichloroethanol, and trichloroethanol glucuronide; these breakdown products are excreted through the kidneys. Most toxic responses occur as a result of industrial exposures. TCE affects principally the central nervous system (CNS). Short exposures result in subjective symptoms such as headache, nausea, and incoordination. Longer exposures may result in CNS depression, hepatorenal failure, and increased cardiac output. Cases of sudden death following TCE exposure are generally attributed to ventricular fibrillation. Current interest in TCE has focused on recent experimental data that implicate TCE as a cause of hepatocellular carcinoma in mice. No epidemiological data are available that demonstrate a similar action in humans. The overall population may be exposed to TCE through household cleaning fluids, decaffeinated coffee, and some spice extracts. The NIOSH recommended standard for TCE is 100 ppm as a time-weighted average for an 8-hr day, with a maximum allowable peak concentration of 150 ppm for 10 min.
Sigma-Aldrich
Trichloroethylene, anhydrous, contains 40 ppm diisopropylamine as stabilizer, ≥99%