- Recombinant human B cell stimulatory factor 2 (BSF-2/IFN-beta 2) regulates beta-fibrinogen and albumin mRNA levels in Fao-9 cells.
Recombinant human B cell stimulatory factor 2 (BSF-2/IFN-beta 2) regulates beta-fibrinogen and albumin mRNA levels in Fao-9 cells.
FEBS letters (1987-08-31)
T Andus, T Geiger, T Hirano, H Northoff, U Ganter, J Bauer, T Kishimoto, P C Heinrich
PMID3305075
ABSTRACT
Conditioned medium from human monocytes contains a partially characterized hepatocyte-stimulating factor that simultaneously elevates the mRNA levels of the acute-phase protein beta-fibrinogen and decreases albumin mRNA in rat hepatoma cells. We demonstrate that recombinant human B-cell stimulatory factor 2, which is identical to interferon-beta 2/26 kDa protein and interleukin-HP1, exhibits the same activity as hepatocyte-stimulating factor. Furthermore, a specific antibody against B-cell stimulatory factor 2 was able to inhibit hepatocyte-stimulating factor in conditioned medium from human monocytes. Our data show that hepatocyte-stimulating factor and B-cell stimulatory factor 2 are functionally and immunologically related proteins.