- Spectrophotometric determination of bismuth(III) with o-hydroxyhydroquinonephthalein in the presence of Brij 58.
Spectrophotometric determination of bismuth(III) with o-hydroxyhydroquinonephthalein in the presence of Brij 58.
A simple and sensitive spectrophotometric determination of bismuth(III) is based on the reaction between bismuth(III) and o-hydroxyhydroquinonephthalein in the presence of Brij 58 in acidic media. The calibration graph is linear over the range 0-3.5 mug/ml bismuth(III) in the final solution, and the apparent molar absorptivity at 520 nm is 9.03 x 10(4) l. mole(-1). cm(-1). The proposed method is 2-10 times more sensitive than other methods, and simpler. It has been applied to the assay of bismuth(III) in pharmaceutical preparations, such as dermatol and bismuth subnitrate, with good results.