- Dysregulation of T cell receptor N-glycosylation: a molecular mechanism involved in ulcerative colitis.
Dysregulation of T cell receptor N-glycosylation: a molecular mechanism involved in ulcerative colitis.
The incidence of inflammatory bowel disease is increasing worldwide and the underlying molecular mechanisms are far from being fully elucidated. Herein, we evaluated the role of N-glycosylation dysregulation in T cells as a key mechanism in the ulcerative colitis (UC) pathogenesis. The evaluation of the branched N-glycosylation levels and profile of intestinal T cell receptor (TCR) were assessed in colonic biopsies from UC patients and healthy controls. Expression alterations of the glycosyltransferase gene MGAT5 were also evaluated. We demonstrated that UC patients exhibit a dysregulation of TCR branched N-glycosylation on lamina propria T lymphocytes. Patients with severe UC showed the most pronounced defect on N-glycan branching in T cells. Moreover, UC patients showed a significant reduction of MGAT5 gene transcription in T lymphocytes. In this study, we disclose for the first time that a deficiency in branched N-glycosylation on TCR due to a reduced MGAT5 gene expression is a new molecular mechanism underlying UC pathogenesis, being a potential novel biomarker with promising clinical and therapeutic applications.