MilliporeSigma
  • A novel system of galangin-potassium permanganate-polyphosphoric acid for the determination of tryptophan and its chemiluminescence mechanism.
A novel system of galangin-potassium permanganate-polyphosphoric acid for the determination of tryptophan and its chemiluminescence mechanism.
Luminescence : the journal of biological and chemical luminescence (2014-10-02)
Li Li, Ruibin Guo, Dongxia Zhang, Xinzhen Du
ABSTRACT
A novel galangin-potassium permanganate (KMnO4)-polyphosphoric acid (PPA) system was found to have an outstanding response to tryptophan (Trp). Trp determination using this KMnO4 -PPA system was enhanced significantly in the presence of galangin. A highly sensitive flow-injection chemiluminescence (CL) method to determine Trp was developed based on the CL reaction of galangin-KMnO4 -Trp in PPA media. The presence of galangin, a member of the flavonol class of flavonoid complexes, greatly increased the luminous intensity of Trp in KMnO4 -PPA systems. Under optimized conditions, Trp was determined in the 0.05-10 µg/mL range, with a detection limit (3σ) of 5.0 × 10(-3)  µg/mL. The relative standard deviation (RSD) was 1.0% for 11 replicate determinations of 1.0 µg/mL Trp. Two synthetic samples were determined selectively with recoveries of 98.4-100.1% in the presence of other amino acids. The possible mechanism is summarized as follows: excited states of Mn(II)(*) and Mn(III(*) types are the main means of generating chemical luminescent species, and Trp concentration and luminescence intensity have a linear relationship, which enables quantitative analysis.