MilliporeSigma
  • Mutations in CNTNAP1 and ADCY6 are responsible for severe arthrogryposis multiplex congenita with axoglial defects.
Mutations in CNTNAP1 and ADCY6 are responsible for severe arthrogryposis multiplex congenita with axoglial defects.
Human molecular genetics (2013-12-10)
Annie Laquérriere, Jérome Maluenda, Adrien Camus, Laura Fontenas, Klaus Dieterich, Flora Nolent, Jié Zhou, Nicole Monnier, Philippe Latour, Damien Gentil, Delphine Héron, Isabelle Desguerres, Pierre Landrieu, Claire Beneteau, Benoit Delaporte, Céline Bellesme, Clarisse Baumann, Yline Capri, Alice Goldenberg, Stanislas Lyonnet, Dominique Bonneau, Brigitte Estournet, Susana Quijano-Roy, Christine Francannet, Sylvie Odent, Marie-Hélène Saint-Frison, Sabine Sigaudy, Dominique Figarella-Branger, Antoinette Gelot, Jean-Marie Mussini, Catherine Lacroix, Valerie Drouin-Garraud, Marie-Claire Malinge, Tania Attié-Bitach, Bettina Bessieres, Maryse Bonniere, Ferechte Encha-Razavi, Anne-Marie Beaufrère, Suonary Khung-Savatovsky, Marie José Perez, Alexandre Vasiljevic, Sandra Mercier, Joelle Roume, Laetitia Trestard, Pascale Saugier-Veber, Marie-Pierre Cordier, Valérie Layet, Marine Legendre, Adeline Vigouroux-Castera, Joel Lunardi, Monica Bayes, Pierre S Jouk, Luc Rigonnot, Michèle Granier, Damien Sternberg, Josiane Warszawski, Ivo Gut, Marie Gonzales, Marcel Tawk, Judith Melki
ABSTRACT
Non-syndromic arthrogryposis multiplex congenita (AMC) is characterized by multiple congenital contractures resulting from reduced fetal mobility. Genetic mapping and whole exome sequencing (WES) were performed in 31 multiplex and/or consanguineous undiagnosed AMC families. Although this approach identified known AMC genes, we here report pathogenic mutations in two new genes. Homozygous frameshift mutations in CNTNAP1 were found in four unrelated families. Patients showed a marked reduction in motor nerve conduction velocity (<10 m/s) and transmission electron microscopy (TEM) of sciatic nerve in the index cases revealed severe abnormalities of both nodes of Ranvier width and myelinated axons. CNTNAP1 encodes CASPR, an essential component of node of Ranvier domains which underlies saltatory conduction of action potentials along the myelinated axons, an important process for neuronal function. A homozygous missense mutation in adenylate cyclase 6 gene (ADCY6) was found in another family characterized by a lack of myelin in the peripheral nervous system (PNS) as determined by TEM. Morpholino knockdown of the zebrafish orthologs led to severe and specific defects in peripheral myelin in spite of the presence of Schwann cells. ADCY6 encodes a protein that belongs to the adenylate cyclase family responsible for the synthesis of cAMP. Elevation of cAMP can mimic axonal contact in vitro and upregulates myelinating signals. Our data indicate an essential and so far unknown role of ADCY6 in PNS myelination likely through the cAMP pathway. Mutations of genes encoding proteins of Ranvier domains or involved in myelination of Schwann cells are responsible for novel and severe human axoglial diseases.