MilliporeSigma
  • Mutations in MRAP, encoding a new interacting partner of the ACTH receptor, cause familial glucocorticoid deficiency type 2.
Mutations in MRAP, encoding a new interacting partner of the ACTH receptor, cause familial glucocorticoid deficiency type 2.
Nature genetics (2005-01-18)
Louise A Metherell, J Paul Chapple, Sadani Cooray, Alessia David, Christian Becker, Franz Rüschendorf, Danielle Naville, Martine Begeot, Bernard Khoo, Peter Nürnberg, Angela Huebner, Michael E Cheetham, Adrian J L Clark
ABSTRACT
Familial glucocorticoid deficiency (FGD), or hereditary unresponsiveness to adrenocorticotropin (ACTH; OMIM 202200), is an autosomal recessive disorder resulting from resistance to the action of ACTH on the adrenal cortex, which stimulates glucocorticoid production. Affected individuals are deficient in cortisol and, if untreated, are likely to succumb to hypoglycemia or overwhelming infection in infancy or childhood. Mutations of the ACTH receptor (melanocortin 2 receptor, MC2R) account for approximately 25% of cases of FGD. FGD without mutations of MC2R is called FGD type 2. Using SNP array genotyping, we mapped a locus involved in FGD type 2 to chromosome 21q22.1. We identified mutations in a gene encoding a 19-kDa single-transmembrane domain protein, now known as melanocortin 2 receptor accessory protein (MRAP). We show that MRAP interacts with MC2R and may have a role in the trafficking of MC2R from the endoplasmic reticulum to the cell surface.