MilliporeSigma
  • Antiretroviral therapy response among HIV-2 infected patients: a systematic review.
Antiretroviral therapy response among HIV-2 infected patients: a systematic review.
BMC infectious diseases (2014-08-27)
Didier K Ekouevi, Boris K Tchounga, Patrick A Coffie, Joseph Tegbe, Alexandra M Anderson, Geoffrey S Gottlieb, Marco Vitoria, François Dabis, Serge P Eholie
ABSTRACT
Few data are available on antiretroviral therapy (ART) response among HIV-2 infected patients. We conducted a systematic review on treatment outcomes among HIV-2 infected patients on ART, focusing on the immunological and virological responses in adults. Data were extracted from articles that were selected after screening of PubMed/MEDLINE up to November 2012 and abstracts of the 1996-2012 international conferences. Observational cohorts, clinical trials and program reports were eligible as long as they reported data on ART response (clinical, immunological or virological) among HIV-2 infected patients. The determinants investigated included patients' demographic characteristics, CD4 cell count at baseline and ART received. Seventeen reports (involving 976 HIV-2 only and 454 HIV1&2 dually reactive patients) were included in the final review, and the analysis presented in this report are related to HIV-2 infected patients only. There was no randomized controlled trial and only two cohorts had enrolled more than 100 HIV-2 only infected patients. The median CD4 count at ART initiation was 165 cells/mm3, [IQR; 137-201] and the median age at ART initiation was 44 years (IQR: 42-48 years). Ten studies included 103 patients treated with three nucleoside reverse transcriptase inhibitors (NRTI). Protease inhibitor (PI) based regimens were reported by 16 studies. Before 2009, the most frequent PIs used were Nelfinavir and Indinavir, whereas it was Lopinavir/ritonavir thereafter. The immunological response at month-12 was reported in six studies and the mean CD4 cell count increase was +118 cells/μL (min-max: 45-200 cells/μL). Overall, clinical and immuno-virologic outcomes in HIV-2 infected individuals treated with ART are suboptimal. There is a need of randomized controlled trials to improve the management and outcomes of people living with HIV-2 infection.
Sigma-Aldrich
Lopinavir, ≥98% (HPLC)
Pricing and availability is not currently available.