MilliporeSigma
  • Biocatalytic synthesis of dihydroxynaphthoic acids by cytochrome P450 CYP199A2.
Biocatalytic synthesis of dihydroxynaphthoic acids by cytochrome P450 CYP199A2.
Bioscience, biotechnology, and biochemistry (2009-12-08)
Toshiki Furuya, Kuniki Kino
ABSTRACT
CYP199A2, a bacterial P450 monooxygenase from Rhodopseudomonas palustris, was found to exhibit oxidation activity towards three hydroxynaphthoic acids. Whole cells of the recombinant Escherichia coli strain expressing CYP199A2 efficiently catalyzed the regioselective oxidation of 1-, 3-, and 6-hydroxy-2-naphthoic acids to produce 1,7-, 3,7-, and 6,7-dihydroxynaphthoic acid respectively. These results suggest that CYP199A2 might be a useful oxidation biocatalyst for the synthesis of dihydroxynaphthoic acids.
Sigma-Aldrich
6-Hydroxy-2-naphthoic acid, 98%
Pricing and availability is not currently available.