MilliporeSigma
  • Rhodium-catalyzed regio- and stereoselective codimerization of alkenes and electron-deficient internal alkynes leading to 1,3-dienes.
Rhodium-catalyzed regio- and stereoselective codimerization of alkenes and electron-deficient internal alkynes leading to 1,3-dienes.
Organic letters (2008-06-12)
Yu Shibata, Masao Hirano, Ken Tanaka
ABSTRACT
A cationic rhodium(I)/H(8)-BINAP complex catalyzes codimerization of alkenes bearing no alpha-hydrogen and electron-deficient internal alkynes, leading to 1,3-dienes in good yields with moderate to excellent regio- and stereoselectivity. The same complex also catalyzes codimerization of an acrylate and phenyl-substituted electron-rich internal alkynes, leading to 1,3-dienes.
Sigma-Aldrich
Ethyl 2-butynoate, 98%
Pricing and availability is not currently available.