MilliporeSigma
  • Controlled template-assisted assembly of plasmid DNA into nanometric particles with high DNA concentration.
Controlled template-assisted assembly of plasmid DNA into nanometric particles with high DNA concentration.
Bioconjugate chemistry (2000-01-19)
M Ouyang, J S Remy, F C Szoka
ABSTRACT
A series of novel cationic detergents that contain cleavable hydrophilic isothiuronium headgroups was synthesized, and their utility in controlled assembly of plasmid DNA into small stable particles with high DNA concentration investigated. The detergents have alkyl chains of C(8)-C(12) and contain hydrophilic isothiuronium headgroups that give relatively high critical micelle concentration (CMC) to the detergents (>10 mM). The isothiuronium group masks a sulfhydryl group on the detergent and can be cleaved in a controlled manner under basic conditions to generate a reactive thiol group. The thiol group can undergo a further reaction after the detergents have accumulated on a DNA template to form a disulfide-linked lipid containing two alkyl chains. The pH-dependent kinetics of cleavage of the isothiuronium group, the CMC of the surfactants, the formation of the complexes, and the transfection efficiency of the DNA complexes have been investigated. Using the C(12) detergent, a approximately 6 kilo-basepair plasmid DNA was compacted into a small particle with an average diameter of around 40 nm with a approximately -13 mV zeta-potential at high DNA concentration (up to 0.3 mg/mL). The compounds were well tolerated in cell culture and showed no cytotoxicity under their CMCs. Under appropriate conditions, the small particle retained transfection activity.