MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • RKIP structure drives its function: a three-state model for regulation of RKIP.
RKIP structure drives its function: a three-state model for regulation of RKIP.
Critical reviews in oncogenesis (2015-01-20)
John J Skinner, Marsha Rich Rosner
ABSTRACT
Raf kinase inhibitory protein (RKIP) is a highly conserved regulator of many signaling networks whose loss or inactivation can lead to a variety of disease states. The multifaceted roles played by RKIP are enabled by an allosteric structure that is controlled through phosphorylation of RKIP and dynamics in the RKIP pocket loop. Perhaps the most striking feature of RKIP is that it can assume multiple functional states. Specifically, phosphorylation redirects RKIP from a state that binds and inhibits Raf-1 to a state that binds and inhibits GRK2. Recent evidence suggests the presence of a third functional state that facilitates RKIP phosphorylation. Here, we present a three-state model to explain the RKIP functional switch and discuss the role of the pocket loop in regulating RKIP activity.