MilliporeSigma
  • Homogeneous liquid-liquid extraction coupled to ion mobility spectrometry for the determination of p-toluidine in water samples.
Homogeneous liquid-liquid extraction coupled to ion mobility spectrometry for the determination of p-toluidine in water samples.
Bulletin of environmental contamination and toxicology (2014-11-28)
Amin Ashori, Ali Sheibani
ABSTRACT
In this research, homogeneous liquid-liquid extraction followed by ion mobility spectrometry (HLLE-IMS) with corona discharge ionization source has been developed for the determination of p-toluidine. The analyte was extracted by single-phase extraction in a ternary solvent system and then the extracted p-toluidine was injected into IMS for analysis. Optimization of different parameters which could influence HLLE-IMS was performed. Under optimum conditions, the dynamic linear range was obtained over 2.0-40.0 ng/mL with R (2) = 0.9966 while relative standard deviation was below 10 %. The limits of detection and quantification were 0.6 and 2.0 ng/mL of p-toluidine, respectively. The proposed method was applied to determine p-toluidine in environmental water samples which resulted in acceptable recoveries of the analyte, ranging from 85.3 %-90 %.
Sigma-Aldrich
p-Toluidine, 99%
Pricing and availability is not currently available.