MilliporeSigma
  • Synthesis and antiviral activities of N-mono- and/or N,N'-di-carbamoyl and acyl derivatives of symmetrical diamines.
Synthesis and antiviral activities of N-mono- and/or N,N'-di-carbamoyl and acyl derivatives of symmetrical diamines.
Chemical & pharmaceutical bulletin (2008-07-02)
Nobuko Mibu, Kazumi Yokomizo, Marumi Oishi, Takeshi Miyata, Kunihiro Sumoto
ABSTRACT
N-carbamoyl and N-acyl diamine derivatives were synthesized from symmetrical diamines by their addition to iso(thio)cyanates, cleavage reaction of acid anhydride, or N-acylation by acyl chloride. (1R,2R)-1,2-Diaminocyclohexane [(1R,2R)-1], meso-1,2-diaminocyclohexane (meso-1), (1R,2R)-1,2-diphenylethylenediamine [(1R,2R)-3], or meso-1,2-diphenylethylenediamine (meso-3) were used as the starting symmetrical diamines. The target compounds synthesized herein were evaluated for antiviral activity with herpes simplex virus type 1 (HSV-1). A few derivatives of 1,2-diaminocyclohexane [(1R,2R)-7aa and cis-7b] with adamantyl group(s) showed significant antiviral activity (EC(50)=16.0, 27.0 microg/ml).
Sigma-Aldrich
meso-1,2-Diphenylethylenediamine, 98%