MilliporeSigma
Association of IL-17RB gene polymorphism with asthma.
Chest (2009-01-02)
Ji-Sun Jung, Byung Lae Park, Hyun Sub Cheong, Joon Seol Bae, Ji-Hye Kim, Hun Soo Chang, TaiYoun Rhim, Jong-Sook Park, An-Soo Jang, Young-Mok Lee, Ki-Up Kim, Soo-Taek Uh, Ju Ock Na, Yong-Hoon Kim, Choon-Sik Park, Hyoung Doo Shin
ABSTRACT
Interleukin (IL)-17E is a member of the IL-17 family, which induces IL-4, IL-5, IL-13, and eotaxin in experimental animals via IL-17 receptor B (IL-17RB). The activation of IL-17RB amplifies allergic-type inflammatory responses by inducing Jun kinase (or JNK), p38 mitogen-activated protein kinase (or MAPK), and nuclear factor-kappaB. We examined the association of polymorphisms in the IL-17RB gene with asthma susceptibility and investigated the effects of those polymorphisms on the transcription of various IL-17RB isoforms. In total, 954 asthmatic patients or 265 healthy control subjects were screened for polymorphisms in IL-17RB by single-base extension. The messenger RNA expression IL-17RB in B-cell lines derived from patients was also measured by reverse transcription-polymerase chain reaction. Direct sequencing of 24 unrelated Korean DNA samples revealed 18 genetic variants, including four insertion/deletions and 14 single-nucleotide polymorphisms (SNPs). Six of the SNPs (-1465G>A, +5661G>A, +6297T>C [Y123Y], +13797C>T, +18661C>T, and +18965G>A) were used to screen a larger group of subjects. Intronic polymorphism +5661G>A was significantly associated with the development of asthma (p = 0.001); moreover, a minor allele of IL-17RB +5661G>A appeared at a lower frequency in the asthmatic patients than in the healthy control subjects (0.13 vs 0.19, respectively). The IL-17RB messenger RNA expression in B cells homozygous for IL-17RB+ 5661GG was significantly higher than that in B cells homozygous for IL-17RB+5661AA (p = 0.002). A rare allele of IL-17RB +5661G>A may have a protective role against the development of asthma via regulation at the level of transcription. The SNPs identified in this study may be used to develop markers to assess the risk of asthma.