MilliporeSigma
  • Palladium-catalyzed, asymmetric Mizoroki-Heck reaction of benzylic electrophiles using phosphoramidites as chiral ligands.
Palladium-catalyzed, asymmetric Mizoroki-Heck reaction of benzylic electrophiles using phosphoramidites as chiral ligands.
Journal of the American Chemical Society (2012-07-04)
Zhigang Yang, Jianrong Zhou
ABSTRACT
We report herein the first examples of asymmetric Mizoroki-Heck reactions using benzyl electrophiles. A new phosphoramidite was identified to be an effective chiral ligand in the palladium-catalyzed reaction. The reaction is compatible with polar functional groups and can be readily scaled up. Several cyclic olefins worked well as olefin components. Thirty-one examples are included.
Sigma-Aldrich
Bis(dibenzylideneacetone)palladium(0)