MilliporeSigma
  • Interferon alpha (IFNα)-induced TRIM22 interrupts HCV replication by ubiquitinating NS5A.
Interferon alpha (IFNα)-induced TRIM22 interrupts HCV replication by ubiquitinating NS5A.
Cellular & molecular immunology (2015-02-17)
Chen Yang, Xinhao Zhao, Dakang Sun, Leilei Yang, Chang Chong, Yu Pan, Xiumei Chi, Yanhang Gao, Moli Wang, Xiaodong Shi, Haibo Sun, Juan Lv, Yuanda Gao, Jin Zhong, Junqi Niu, Bing Sun
ABSTRACT
TRIM22, a tripartite-motif (TRIM) protein, is upregulated upon interferon alpha (IFNα) administration to hepatitis C virus (HCV)-infected patients. However, the physiological role of TRIM22 upregulation remains unclear. Here, we describe a potential antiviral function of TRIM22's targeting of the HCV NS5A protein. NS5A is important for HCV replication and for resistance to IFNα therapy. During the first 24 h following the initiation of IFNα treatment, upregulation of TRIM22 in the peripheral blood mononuclear cells (PBMCs) of HCV patients correlated with a decrease in viral titer. This phenomenon was confirmed in the hepatocyte-derived cell line Huh-7, which is highly permissive for HCV infection. TRIM22 over-expression inhibited HCV replication, and Small interfering RNA (siRNA)-mediated knockdown of TRIM22 diminished IFNα-induced anti-HCV function. Furthermore, we determined that TRIM22 ubiquitinates NS5A in a concentration-dependent manner. In summary, our results suggest that TRIM22 upregulation is associated with HCV decline during IFNα treatment and plays an important role in controlling HCV replication in vitro.