MilliporeSigma
  • A new automated method for phenotyping arylesterase (EC 3.1.1.2) based upon inhibition of enzymatic hydrolysis of 4-nitrophenyl acetate by phenyl acetate.
A new automated method for phenotyping arylesterase (EC 3.1.1.2) based upon inhibition of enzymatic hydrolysis of 4-nitrophenyl acetate by phenyl acetate.
European journal of clinical chemistry and clinical biochemistry : journal of the Forum of European Clinical Chemistry Societies (1992-07-01)
L Haagen, A Brock
ABSTRACT
A new method for phenotyping human serum arylesterase (EC 3.1.1.2) is described and evaluated. The aromatic esters, phenyl acetate and 4-nitrophenyl acetate, were compared as substrates for spectrophotometric measurement of arylesterase activity. A method for arylesterase phenotyping, based upon inhibition of the enzymatic hydrolysis of 4-nitrophenyl acetate by phenyl acetate, was developed. The method was applied to serum samples from 158 blood donors and showed a distinct separation of the three phenotypes defined by a reference method based on the ratio of paraoxonase activity to arylesterase activity using paraoxon and phenyl acetate as substrates. The method was adapted to a Cobas-Fara centrifugal analyser.
Sigma-Aldrich
Phenyl acetate, 99%